Search
| 카탈로그 번호 | 색상 | 오토클레이브 가능 | 제품 사이즈 |
|---|---|---|---|
| 525-A-Q | Amber | Autoclavable | Case of 10 |
| 525-B-Q | Blue | Autoclavable | Case of 5000 |
| 525-B-QPK 525-B-Q으로도 사용됨 | Blue | Autoclavable | Pack of 500 |
| 525-G-Q | Green | Autoclavable | Case of 5000 |
| 525-G-QPK 525-G-Q으로도 사용됨 | Green | Autoclavable | Pack of 500 |
| 525-MIXED-Q | Assorted | Autoclavable | Case of 10 |
| 525-O-Q | Orange | Autoclavable | Case of 10 |
| 525-R-Q | Red | Autoclavable | Case of 5000 |
| 525-R-QPK 525-R-Q으로도 사용됨 | Red | Autoclavable | Pack of 500 |
| 525-V-Q | Violet | Autoclavable | Case of 10 |
| 525-Y-Q | Yellow | Autoclavable | Case of 10 |
Thermo Scientific™ QSP™ Screw Cap Microcentrifuge Tube Caps with O-rings, Amber, packaged bulk bag non-sterile with clean claims; RNase/DNase, DNA, pyrogen and endotoxin free.
The robust design and material selection are great for use with biologics, sample preparation, storage and transportation.