Nalgene™ Replacement Plates for Vacuum Chambers, polycarbonate
Nalgene™ Replacement Plates for Vacuum Chambers, polycarbonate
Nalgene™ Replacement Plates for Vacuum Chambers, polycarbonate
Nalgene™ Replacement Plates for Vacuum Chambers, polycarbonate
Thermo Scientific™
Nalgene™ Replacement Plates for Vacuum Chambers, polycarbonate
Use these replacement plates for select vacuum chambers and bell jars. Plates are molded of white polypropylene or polycarbonate, with neoprene gaskets.
Have Questions?
카탈로그 번호유형
5306-0130PKVacuum Plate
카탈로그 번호 5306-0130PK
제품 가격(KRW)
105,000
Each
유형:
Vacuum Plate
제품 가격(KRW)
105,000
Each
Use Thermo Scientific™ Nalgene™ Replacement Plate for Vacuum Chambers with Nalgene Vacuum Chambers (5305-series), Nalgene Replacement Bell Jars (DS5320 Series), or with glass bell jars up to 12.5 in. (31.75cm) in diameter. Plate is molded of white polycarbonate, with neoprene gaskets.
Features:
  • For use with Nalgene Vacuum Chambers (5305-series), Nalgene Replacement Bell Jars (DS5320 Series), or with glass bell jars up to 12.5 in. (31.75cm) in diameter
  • Plates are 1.06 in. H (2.7cm)
  • Supplied with 0.09 in. thick (2.4mm) neoprene gaskets
  • Tubing adapter conveniently located on rim of plate, fits 0.25″ I.D. (6mm) tubing
Ordering Information:
Contact your customer service representative for ordering information on replacement bell jars and gasket kit.
Please recycle the corrugate cartons in which your Nalgene products are packaged.
사양
용도(장비)Nalgene™ Vacuum Chambers
재질(플레이트)Polycarbonate
케이스당 수4
팩당 수1
외경(영국식 단위) 개스킷8, 11, 13 in.
외경(영국식 단위) 플레이트13.5 in.
외경(미터법) 개스킷20, 28, 33 cm
외경(미터법) 플레이트343 mm
두께(영국식 단위)1-1/16 in.
두께(미터법)27 mm
색상White
물질Polycarbonate (Plate); Neoprene (Gaskets)
유형Vacuum Plate
Unit SizeEach