Thermo Scientific™

DyLight™ 780-B2 NHS Ester

Catalog number:  53065

 Related applications: Protein Labeling & Crosslinking

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific DyLight near-infrared specialty dyes, comparable to Alexa Fluor and IRDye NIR dyes, can be used to label antibodies, peptides, and other proteins at primary amines. DyLight 780-B2 dye has a structure based on the benzopyrillium core, with 2 sulfonates. It has excitation and emission peaks at 784 and 796 nm, respectively (in ethanol).

General characteristics of DyLight near-infrared emitting specialty dyes:

Large selection—the largest family of dyes available for NIR fluorescence applications
NHS ester reactive group—allows immediate labeling of antibodies, proteins, peptides and other amine-containing molecules through amide bond formation
Broad spectrum of water solubilities—choose from hydrophilic to hydrophobic dyes to optimize the right dye label for the best performance in a given application
NIR dyes avoid background interference—DyLight NIR Dyes avoid fluorescence interference or quenching effects from biomolecules present in samples
Excellent signal penetration through cells and tissues—DyLight NIR Dyes provide the optimal window for excitation and emission for in vivo imaging applications

DyLight NIR Dyes are a family of labeling agents that can be used for bright fluorescence detection in cell-based imaging or in vivo imaging applications. NIR dyes can be selected based upon their characteristic excitation and emission properties or relative hydrophilicity and hydrophobicity attributes. Dyes that contain a greater number of negatively charged sulfonates generally will have greater water solubility than dyes with fewer sulfonates. More hydrophobic dyes often provide better cell penetrating ability in vivo, while more hydrophilic dyes have less nonspecific binding potential. Each dye contains an amine-reactive NHS ester for simple modification of antibodies, proteins, peptides or other biomolecules through amide bond formation. NIR dyes are best for imaging through tissues and away from indigenous fluorescent biomolecule interference or quenching. DyLight Near Infrared Dyes represent the largest selection of fluorescent labels that are commercially available.

Criteria to consider when choosing a DyLight NIR Specialty Dye
• Excitation and emission wavelengths—choose the best dye to match the excitation and emission capabilities of your instrument
• Water solubility—choose a DyLight NIR Dye based on its relative hydrophilicity, which directly correlates to the number of negatively-charged sulfonates it has on its core structure. More hydrophilic dyes are best at maintaining water solubility of a labeled antibody and limiting the nonspecific binding of the conjugate. More hydrophobic dyes often are best at penetrating tissues and cell membranes in vivo, meaning that dyes with fewer sulfonates may work best for some applications.
• DyLight Dye selection—the broad selection of NIR dyes allows a number of candidate dyes to be tested in a given application for optimal performance.

Applications:
In vivo or ex vivo imaging
• Tumor imaging with labeled peptides
• NIR fluorescence (NIRF) imaging of labeled silica nanoparticles
• NIR in vitro imaging and characterization
• Determination of thermal stability
• Cytotoxicity assays
• Molecular imaging
• UV-VIS-NIR spectroscopy
• Fluorescence correlation spectroscopy
• MRI applications
• DNA sequencing
• Primer labeling for PCR
• 2-D gel electrophoresis
• Flow cytometry/fluorescence-activated cell sorting (FACS)
• Laser scanning confocal microscopy

Related Products
DyLight™ 780-B1 NHS Ester
DyLight™ 780-B3 NHS Ester
DyLight™ 830-B2 NHS Ester
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: DyLight™ Fluor (Specialty)
Chemical Reactivity: Amine
Excitation⁄Emission (nm): 784⁄796
Product Line: DyLight™
Product Size: 1 mg
Solubility: DMF (Dimethylformamide)

Contents & storage

Store desiccated at -20°C. Shipped at ambient temperature.

Documents

Manuals & protocols