SeCore® B Locus Sequencing Kit (RUO)
Catalog number: 5310925
Invitrogen™  Related applications: Diagnostic Testing
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The Class I SeCore sequencing strategy begins with a multiplex PCR. Two fragments are amplified; one amplicon encompassing exon 2 and 3 and the other encompassing exon 4. The sequencing mixes are designed for bi-directional sequencing of each exon. The B Locus kit includes two amplification mixes, each specific to a set of alleles. Heterozygous sample alleles are often amplified by different mixes and therefore are separated by this initial PCR. The resulting typing data for each separated allele is a greatly simplified homozygous electropherogram, eliminating the challenge of interpreting polymorphic positions. This strategy reduces the cis/trans ambiguities for B locus by 45%. The Class I Z GSSP Kits are designed to resolve ambiguous allele combinations encountered with the SeCore A, B and C Locus Sequencing Kits. Additional amplification is not necessary; the original amplicon serves as the GSSP template. To identify the GSSP Kit that will resolve your ambiguity click on the link "SeCore GSSP Selection Table".
Contents & storage
Store in freezer (-5 to -30°C).
Documents
Manuals & protocols