SeCore® DQB1 Locus 2AMP
Catalog number: 5341025D
Invitrogen™  Related applications: Diagnostic Testing
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
Two SeCore DRB sequencing strategies are available to best suit your resolution and throughput needs; DRB1 Locus Sequencing Kit (Single Amplification System) and DRB1, DRB3, 4 and 5 Sequencing Kit (Group Specific Set). Both formats are designed for bi-directional sequencing of exon 2 and include a Codon 86 Sequencing Mix.
Strategy 1:
The SeCore DRB1 Locus Sequencing Kit (Single Amplification System) begins with only one PCR per sample. This method eliminates the need for a confirmatory agarose gel and offers the user the most high throughput route for DRB1 sequencing.
Stategy 2:
The SeCore DRB1, DRB3, 4 and 5 Sequencing Kit (Group Specific Set) provides eleven group specific amplification mixes. All eleven amplification mixes can be used for each sample, or the user can select only the amplification mixes necessary for each sample based on previous low resolution data. The resulting typing data for each separated allele is a greatly simplified homozygous electropherogram, eliminating the challenge of interpreting polymorphic positions. This strategy reduces the cis/trans ambiguities for DRB locus by 98%.
Additional Resolution: DRB1 ambiguities can be further resolved by SeCore Group Specific Sequencing Products (GSSP). The amplicon from either SeCore DRB core kits serves as the template. GSS-Products contain a single group specific unidirectional sequencing mix and precipitation buffer. Sequencing parameters are identical to the other core SeCore products. All GSS-Products are sold Research Use Only (RUO).
The SeCore DQB1 Locus Sequencing Kit amplifies and bidirectionally sequences exon 2 and exon 3.
The SeCore DPB1 Locus Sequencing Kit includes four amplification mixes, each specific to a set of alleles. Heterozygote sample alleles are often amplified by different mixes and therefore are separated by this initial PCR. The resulting typing data for each separated allele is a greatly simplified electropherogram, eliminating the challenge of interpreting polymorphic positions. This strategy reduces the cis/trans DPB1 Locus ambiguities by 70%.
The SeCore® HLA Sequencing System is intended for the identification and definition of Class I and II Human Leukocyte Antigens (HLA) and is for use in HLA typing. The SeCore® HLA Sequencing System provides human histocompatibility information of HLA Class I (A, B, and C) and Class II (DPB1, DQB1, and DR) loci using genomic DNA isolated from whole blood specimens. HLA typing using the SeCore® Sequencing Kits must be performed in the presence of an HLA Lab Director, Technical Supervisor and/or general Supervisor following accepted laboratory accreditation standards (ASHI). These products are for professional use only. This test must not be used as the sole basis for making clinical decisions. For the intended use of individual system components, refer to the documentation provided with the component.
“This reagent is classified as a medical device in several countries. If it is not regulated as a medical device in specific region, it should be used for laboratory use only.”
Contents & storage
Store in freezer (-5 to -30°C).
Documents
Manuals & protocols
Recommended products