uTYPE® Dx SBT Analysis Software
Catalog number: 539993
Invitrogen™  Related applications: Diagnostic Testing
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
uTYPE® Dx SBT Analysis Software and support manual is provided as a CD. It is used to produce in vitro diagnostic human leukocyte antigen (HLA) typing results from data generated by the 3500 Dx Genetic Analyzer CS2 instruments.
Features include:
• Processes .ab1 files from the 3500 Dx Genetic Analyzer CS2 instruments (8 and 24 capillary)
• Used to produce in vitro diagnostic (IVD)-labeled results
• Imports the uTYPE® Dx Allele Library, an allele variant sequence database
This software compares data produced by the genetic analyzer to the allele library (variant database) and outputs sample HLA typing results. Typing can be done for the following loci: A, B, C, DRB1, DRB Group, DQB1, and DPB1. It is a required part of the diagnostic HLA workflow. Included in this workflow are the following: SeCore®HLA typing kits, the Veriti™ Dx 96-well Thermal Cycler, the 3500 Dx Genetic Analyzer CS2 instruments (8 and 24 capillary), the uTYPE® Dx SBT Analysis Software, and the uTYPE® Dx Allele Library. All Dx components must be used to produce HLA IVD typing results.
The uTYPE® Dx v1.0 HLA Sequence Analysis Software is intended to interpret and match sequencing data generated on the Applied Biosystems® 3500 Dx /3500 xL Dx Genetic Analyzer CS2 with the 3500 Dx Series Data Collection Software using HLA sequencing assay kits to known HLA type sequences.
The SeCore® HLA Sequencing System is intended for the identification and definition of Class I and II Human Leukocyte Antigens (HLA) for use in HLA typing. The SeCore® HLA Sequencing System provides human histocompatibility information of HLA Class I (A, B, and C) and Class II (DPB1, DQB1, and DR) loci using genomic DNA isolated from whole blood specimens.
Contents & storage
One CD. Store at room temperature.
Documents
Manuals & protocols