Thermo Scientific™ QSP™ Plug Cap Tube Closure
Thermo Scientific™

Thermo Scientific™ QSP™ Plug Cap Tube Closure

Thermo Scientific™ QSP™ Plug Caps make a snug fit, leak resistant closure for 5ml (12x75), 12mm or 13mm tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
540-B-QBlue
540-G-QGreen
540-N-QNatural
540-QWhite
540-R-QRed
540-Y-QYellow
Catalog number 540-B-Q
Price (MXN)
-
Color:
Blue

Thermo Scientific™ QSP™ Plug Cap Tube Closure fits 5ML, 12x75 tubes, 12mm and 13mm tubes, knurled cap helps with grip and the cap collar has ribs securing cap in the tube, Blue color. Non-sterile clean claims; Rnase/Dnase, DNA, pyrogen and endotoxin free.

  • Non-autoclavable low density polyethylene material
  • Colored options
  • Plug closure leak resistant
Specifications
Clean ClaimsNon-Sterile free of: Rnase/Dnase, DNA, pyrogen and endotoxin
Closure Size5 mL
For Use With (Equipment)5 mL (12x75), 12 mm or 13 mm Tubes
AutoclavableNot Autoclavable
ColorBlue
MaterialLDPE
SterilityNon-sterile
Unit SizeCase of 5000