Thermo Scientific™ QSP™ Plug Cap Tube Closure
Thermo Scientific™
Thermo Scientific™ QSP™ Plug Cap Tube Closure
Thermo Scientific™ QSP™ Plug Caps make a snug fit, leak resistant closure for 5ml (12x75), 12mm or 13mm tubes.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoColor
540-QBlanco
540-B-QAzul
540-G-QVerde
540-N-QNatural
540-R-QRojo
540-Y-QAmarillo
Número de catálogo 540-Q
Precio (CLP)
-
Color:
Blanco
Thermo Scientific™ QSP™ Plug Cap Tube Closure fits 5ML, 12x75 tubes, 12mm and 13mm tubes, knurled cap helps with grip and the cap collar has ribs securing cap in the tube, Blue color. Non-sterile clean claims; Rnase/Dnase, DNA, pyrogen and endotoxin free.
  • Non-autoclavable low density polyethylene material
  • Colored options
  • Plug closure leak resistant
Especificaciones
Declaraciones de limpiezaNon-Sterile free of: Rnase/Dnase, DNA, pyrogen and endotoxin
Tamaño del tapón5 mL
Para utilizar con (equipo)5 mL (12x75), 12 mm or 13 mm Tubes
Esterilizables en autoclaveNot Autoclavable
ColorBlanco
MaterialLDPE
EsterilidadNo estéril
Unit SizeCase of 5000