KingFisher Apex Purification Systems
Thermo Scientific™

KingFisher Apex Purification Systems

The KingFisher Apex Purification System is a versatile and user-friendly instrument that uses magnetic particle processing to automate the purification or isolation of DNA, RNA, proteins, and cells from virtually any source.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
540093096 DW head
540091096 PCR Head
A4694196 PCR head, Ext Warranty
540092096 Combi head
A4694396 Combi head, Ext Warranty
A4694496 DW head, Ext Warranty
540094024 Combi head
A4694224 Combi Head, Ext Warranty
Catalog number 5400930
Price (TWD)
-
Product Type:
96 DW head

The KingFisher Apex Purification System is a versatile and user-friendly instrument that uses magnetic particle processing to automate the purification or isolation of DNA, RNA, proteins, and cells from virtually any source. It moves magnetic beads, not liquids, through the main steps of sample preparation: bind, wash, and elute. This process provides excellent quality, reproducible results, and reduces hands-on time for sample processing.

  • User-friendly high throughput sample purification system for automating DNA/RNA, protein and cell purification
  • Provides versatile and high-speed purification with reproducible results
  • Easy to use interface with touch screen empowering the user to control all details of the application and purification process
  • BindIx PC Software for added convenience
  • Extensive selection of validated protocols for different type of applications
  • Cloud library for quick protocol retrieval
Specifications
Collection Efficiency of Particles>98% with 2.8 μm particle size
Cooling TemperatureDown to +4°C
Depth (English)22 in.
Depth (Metric)56 cm
DisplayTouch screen (10 in.)
For Use With (Application)Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics
Heating Temperature+4°C above ambient temperature up to +100°C
Height (English)18 in.
Height (Metric)46 cm
Magnetic Rods96 in one frame, interchangeable
Particle Size (Metric)ca. >1 μm
Plate Type96 KF, 96 DW
Samples per Run96
Type of Magnet Head96 in one frame, interchangeable
UV Exposure TimeMax 23 hr. 59 min.
UV Lamp2 pcs
Weight (English)123 lb.
Weight (Metric)56 kg
Width (English)31 in.
Width (Metric)79 cm
Capacity96 Samples
Format96-well
Plates per Deck8
Product Type96 DW head
Sample TypePlasma, Blood, Urine, Saliva, Cell Homogenate, Tissue Homogenate
Suitable Downstream AnalysisPCR/qPCR, Sequencing, Mass Spec
Volume (Metric) Processing96 KF plate 50 to 150 μL, 96 DW plate 50 to 1000 μL
Unit SizeEach

Frequently asked questions (FAQs)

What should I do if the tip comb does not load correctly onto my Thermo Scientific KingFisher system?

If the tip does not load correctly onto a Thermo Scientific KingFisher magnetic head, we recommend that you try manually flexing the tips both lengthwise and widthwise to level the tip comb. A video of flexing the tip comb can be found here: https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/automated-purification-extraction/kingfisher-systems/resources.html. The video is under "KingFisher Flex" and titled "How to install the magnet head and check the tip comb." Flexing of the tip comb is demonstrated at timepoint 1:35. If that does not work, try a different box or lot of tips. This applies to all KingFisher instruments.