Thermo Scientific™
Vytac™ Formalin Disposal System
Related applications:
Clinical
Safely dispose of both 10% buffered and unbuffered formalin, including zinc formalin, using the Thermo Scientific™ Vytac™ Formalin Disposal System. This simple, economical system eliminates the need for expensive waste hauling and onsite permits. 
Back to top
Disposal Solution:
  • Breaks down formalin into inert components
  • Neutralizes four parts formalin waste per one part Vytac
  • When used as directed, neutralizes more than 99% of the waste formalin collected
Note: Vytac GNF will neutralize gluteraldehyde