AllSet+™ Gold SSP B*44/45 High-Resolution Kit

Catalog number: 54150D

 Related applications: Diagnostic Testing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Straight from genomic DNA to high resolution results, no need for initial low-resolution typing

• Allele level typing with a single tray
• Save time and QC costs
• Ready to use, dry pre-dispensed primer mixes
• Dried, pre-aliquoted primer mixes
• Includes pre-aliquoted amplification buffer
• Compatible with UniMatch® Plus Analysis software
• Allele-updating available via the web

Contents & storage

Storage: +4°C to -20°C
Shipping: Room temperature or wet ice

Documents

Manuals & protocols