AllSet+™ Gold DQB1*05 High-Resolution SSP Kit

Catalog number: 542150D

 Related applications: Diagnostic Testing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

For use when high resolution typing of HLA Class II is required and low resolution results are known.

Provides allele-level typing results with minimal ambiguities.

Benefits:

• Easy technique and interpretation
• Dried, pre-dispensed primer mixes for easy storage and use
• Pre-aliquotted amplification buffer saves time
• Negative contamination well with each test ensures quality
• Color coded primer mix to confirm orientation
• Frequently updated for new alleles
• Superior technical and customer service
• Convenient shipping and storage conditions

Compatible with UniMatch® Plus software, and includes SCORE file, for easy interpretation.

Contents & storage

Storage: +4°C to -20°C
Shelf Life: 18 months
Shipping: Room temperature or wet ice

Documents

Manuals & protocols