AllSet+™ Gold HLA C Low-Resolution SSP Kit

Catalog number: 54330D

Invitrogen™  Related applications: Diagnostic Testing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


• Provides low to intermediate HLA typing results
• Products offered in a variety of formats, including combo kits
• Quick turn-around-time makes this technology ideal for on-call typing
• Pre-dispensed dry primer mixes
• Includes pre-aliquotted amplification buffer, saving time and labor
• Compatible with UniMatch® Plus Analysis software and includes Score file

Contents & storage

Storage: +4°C to -20°C
Shelf Life: 18 months
Shipping: Room temperature or wet ice


Manuals & protocols