Electrochemical Detector Cells
Actual product may vary
Thermo Scientific™
Electrochemical Detector Cells
Choose from an extensive selection of electrochemical cells which have been optimized for a variety of critical applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
55-0417Preanalytical, Flow-Through
55-0685Coulometric, Flow-Through
Catalog number 55-0417
Price (MXN)
-
Type:
Preanalytical, Flow-Through
Choose from this selection of electrochemical cells which have been optimized for a variety of critical applications. Thermo Scientific™ Dionex™ Electrochemical Detector Cells for LC are unique in their configuration and performance. Our advanced sensor designs assure long-lasting, trouble-free performance.
For Use with Thermo Scientific Dionex Electrochemical Detectors
These Electrochemical Cells are recommended for use with the following detectors, which are compatible with all Thermo Scientific Dionex UHPLC+ systems:
  • Dionex™ Coulochem™ III Electrochemical Detector
  • Dionex™ CoulArray™ Coulometric Array Detector
Specifications
DescriptionGuard Cell, Model 5020
Detection ModeDC
For Use With (Equipment)Dionex Coulochem Series Detectors
No. of Channels0
Reference ElectrodePd
Working ElectrodesPG
Detector TypeElectrochemical (ECD)
TypePreanalytical, Flow-Through
Unit SizeEach