Axiom™ Strawberry Genotyping Array
Applied Biosystems™

Axiom™ Strawberry Genotyping Array

Axiom Strawberry Genotyping Array was designed through the Affymetrix Expert Design Program in collaboration with the International RosBREED SNP consortiumRead more
Have Questions?
Catalog NumberQuantity
5504661 plate
Catalog number 550466
Price (MXN)
-
Quantity:
1 plate
Axiom Strawberry Genotyping Array was designed through the Affymetrix Expert Design Program in collaboration with the International RosBREED SNP consortium that spans multiple nations, states, and institutions dedicated to genetic improvement of rosaceous crops.* The array offers genome-wide coverage of polymorphic SNPs across the cultivated garden strawberry hybrid (Fragaria ananassa).

Features of Axiom Strawberry Genotyping Array:
• Comprehensive content: 95,062 SNPs and indels derived from octoploid and diploid cultivars, including:
   - 1,761 multi-allelic SNPs
   - 3,751 SNPs derived from diploid cultivars; the remaining markers are derived from octoploid cultivars
• Multiple cultivars represented: SNP discovery was facilitated with a diverse worldwide breeding germplasm panel that included:    - Holiday, Korona, and F1 seedlings from the cross Holiday x Korona
   - Two likely diploid progenitors, Fragaria mandschurica, F. iinumae
   - One known diploid F. vesca

Applications of Axiom Strawberry Genotyping Array:
•Complex trait research
   - QTL discovery
   - Identify traits of economic significance
•Molecular breeding
   - Genome-wide scanning
   - Marker-assisted breeding
•Release of new cultivars
   - Accelerate and increase efficiency of cultivar development
   - Identify the presence of important markers in existing strawberry germplasm
   - Use genetic information to identify germplasm that will perform well in specific growing regions

Notes about the data analysis: Axiom Strawberry Genotyping Array requires advanced analysis. Genotyping many different varieties together may create multiple clusters during the analysis that interfere with each other and/or blend the clusters together. Genotype calls are not provided for multi-allelic SNPs.

*Expert Design Arrays are designed by key opinion leaders in the plant and animal genomics community and made available to the public for the purpose of advancing research and breeding programs. These arrays contain genetic markers and annotations that are provided by the original customer who designed the array and are therefore not updated by Affymetrix in the same manner as other catalog products. Through the program, Affymetrix continually provides agrigenomics researchers access to genotyping arrays that have been utilized by their peers in the research community.

Required Products
Axiom Analysis Suite
GeneChip Command Console Software
GeneTitan Multi-Channel Instrument
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Microarray Analysis
Quantity1 plate
ArrayGenotyping
Format96-array Plate
Number of Arrays96 arrays
SpeciesStrawberry
Unit Size1 plate

Frequently asked questions (FAQs)

What is the starting amount of DNA required for Axiom arrays?

All Axiom arrays (except for Axiom Genome-Wide Pan-African Array Set) require a total of 200 ng DNA. The Axiom Genome-Wide Pan-African Array Set requires a total of 300 ng DNA, or 100 ng DNA per array (there are 3 arrays).

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What are the sample requirements for Axiom array processing at the Thermo Fisher Scientific Microarray Research Services Laboratory (MRSL)?

For information regarding our in-house MRSL requirements, please contact us via phone at 888-362-2447 (in North America) or via email at BioinformaticsServices@thermofisher.com.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.