Axiom™ Salmon Genotyping Array
Applied Biosystems™

Axiom™ Salmon Genotyping Array

Axiom Salmon Genotyping Array for Atlantic salmon (Salmo salar) was designed under the Affymetrix Expert Design Program in collaboration withRead more
Have Questions?
Catalog NumberQuantity
5505401 plate
Catalog number 550540
Price (USD)
-
Quantity:
1 plate
Axiom Salmon Genotyping Array for Atlantic salmon (Salmo salar) was designed under the Affymetrix Expert Design Program in collaboration with The Roslin Institute (University of Edinburgh), Edinburgh Genomics, Landcatch Natural Selection Ltd., University of Stirling, and University of Glasgow in the United Kingdom.*

The array interrogates more than 130,000 polymorphic variants that were discovered through three different sequencing methodologies. Axiom Salmon Genotyping Array provides extensive information on polymorphisms across the salmon genome.

The high marker density on Axiom Salmon Genotyping Array ensures a broad coverage of the salmon genome and avoids underrepresentation of important regions of the salmon genome. This is a significant improvement over existing Atlantic salmon genotyping tools.

Features of Axiom Salmon Genotyping Array:
Diverse content – More than 130,000 SNPs evenly spaced across the genome with preference given to transcribed regions. The array includes 4,714 markers from public sources and 87 Y chromosome-specific probes for accurate gender determination.
Multiple breeds represented – Farmed Scottish and Norwegian and wild Scottish, Irish, Norwegian, and Spanish populations

Applications of Axiom Salmon Genotyping Array:
• Complex trait research
   - GWAS
   - Fine mapping of QTL affecting economically important traits in salmonids
• Molecular breeding
   - Association mapping and genomic selection
   - Improved accuracy in aquaculture breeding programs
   - Gender determination via Y chromosome-specific probes
• Population and evolutionary genetics
   - Distinguish between fish of different origins
   - Differentiate farmed and wild populations

*Expert Design Arrays are designed by key opinion leaders in the plant and animal genomics community and made available to the public for the purpose of advancing research and breeding programs. These arrays contain genetic markers and annotations that are provided by the original customer who designed the array and are therefore not updated by Affymetrix in the same manner as other catalog products. Through the program, Affymetrix continually provides agrigenomics researchers access to genotyping arrays that have been utilized by their peers in the research community.

Required Products
Axiom Analysis Suite
GeneChip Command Console Software
GeneTitan Multi-Channel Instrument
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Microarray Analysis
Quantity1 plate
ArrayGenotyping
Format96-array Plate
Number of Arrays96 arrays
SpeciesSalmon
Unit Size1 plate

Frequently asked questions (FAQs)

What is the starting amount of DNA required for Axiom arrays?

All Axiom arrays (except for Axiom Genome-Wide Pan-African Array Set) require a total of 200 ng DNA. The Axiom Genome-Wide Pan-African Array Set requires a total of 300 ng DNA, or 100 ng DNA per array (there are 3 arrays).

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What are the sample requirements for Axiom array processing at the Thermo Fisher Scientific Microarray Research Services Laboratory (MRSL)?

For information regarding our in-house MRSL requirements, please contact us via phone at 888-362-2447 (in North America) or via email at BioinformaticsServices@thermofisher.com.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.