Axiom™ Porcine Breeders Array, mini-96 format
Applied Biosystems™

Axiom™ Porcine Breeders Array, mini-96 format

The Applied Biosystems Axiom Porcine Breeders Array includes markers focused on genomic selection of commercial porcine breeds, including Large White,Más información
Have Questions?
Número de catálogoCantidad
5508551 placa
Número de catálogo 550855
Precio (MXN)
-
Cantidad:
1 placa
The Applied Biosystems Axiom Porcine Breeders Array includes markers focused on genomic selection of commercial porcine breeds, including Large White, Landrace, Duroc, and Pietrain. Core markers for the array were selected from markers on the Axiom Porcine High-Density Array that were evaluated using samples from multiple commercial and domestic breeds, including European wild boars. The inclusion of the most informative and well-performing markers from existing porcine arrays ensures that this array can be used for comparison with data generated by previous studies. With the availability of the Axiom Long Format Export Tool, which converts Axiom array data into long format, the array also provides the ability to continue existing projects with no change to the current bioinformatics infrastructure and pipeline.

Features include:
• 55,150 markers for genomic and trait selection
• 42 trait-specific markers selected by United States Department of Agriculture
• 64 parentage markers, including ISAG core markers
• Evenly distributed SNPs across the porcine genome
• Marker annotations aligned to Sscrofa10.2/susScr3 genome and dbSNP 148
• Targeted for commercial breeds including Large White, Landrace, Duroc, and Pietrain

Applications
• Parentage analysis
• Identification of economically important traits
• Genetic improvement of pure lines

Required products
GeneChip Command Console Software
GeneTitan Multi-Channel Instrument
Axiom Analysis Suite Software

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.

Especificaciones
Para utilizar con (aplicación)Microarray Analysis
Línea de productosAxiom
Cantidad1 placa
matriz, red, conjuntoDNA
Formato96-array Plate
Número de arrays96 matrices
Unit Size1 plate
Contenido y almacenamiento
One plate containing 96 arrays