Axiom™ Bovine Genotyping v3 Array (384HT format)
Axiom™ Bovine Genotyping v3 Array (384HT format)
Applied Biosystems™

Axiom™ Bovine Genotyping v3 Array (384HT format)

The Applied Biosystems Axiom Bovine Genotyping v3 Array has >63,000 markers and is designed for genomic and trait selection andRead more
Have Questions?
Catalog NumberQuantity
5510891 plate
Catalog number 551089
Price (USD)
-
Request A Quote
Quantity:
1 plate
The Applied Biosystems Axiom Bovine Genotyping v3 Array has >63,000 markers and is designed for genomic and trait selection and parentage verification of cattle. The single-nucleotide polymorphisms (SNPs) on the array were selected based on their position in the bovine genome, ensuring even spacing. These include VIP trait-associated SNPs that are publically available, ISAG core parentage markers, and 44,000 markers from the Council on Dairy Cattle Breeding (CDCB). SNPs optimized for STR imputation (1) are also included in the array content. This array is also available in mini-96 format.

Traits associated with fertility are on the Y-chromosome, so over 500 markers on the Y-chromosome are included in this array. The array also contains >10,000 Bos indicus markers of approximate equal coverage across the Bos indicus genome to avoid gaps.

Applications
• Parentage analysis
• Identification of economically important traits
• Dairy evaluation
• Genetic improvement of pure lines

Required products
Axiom Long Format Export Tool
Command Console Software
GeneTitan Multi-Channel Instrument
Axiom Analysis Suite Software

Reference
1. McClure MC, Sonstegard TS, Wiggans GR, et al. Imputation of microsatellite alleles from dense SNP genotypes for parentage verification across multiple Bos taurus and Bos indicus breeds. Front Genet. 2013;4:176.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Microarray Analysis
Quantity1 plate
Shipping ConditionRoom temperature
Format384 HT-array Plate
Number of Arrays384 arrays
SpeciesBovine
Unit Size1 plate
Contents & Storage
One plate containing 384 arrays, Store in refrigerator (2-8°C)