Thermo Scientific™
Richard-Allan Scientific™ Cal-Rite™ Decalcifying/Fixation Solution
Related applications:
Anatomical Pathology
Streamline processes with Thermo Scientific™ Richard-Allan Scientific™ Cal-Rite™ Decalcifying/Fixation Solution, a mixture of formic acid and formaldehyde which allows simultaneous decalcification and fixation.
Back to top
  • Beneficial for conventional iron staining
  • Preserves nuclear chromatin staining, even after prolonged exposure
  • Eliminates poor hematoxylin and eosin staining caused by over-decalcification and/or exposure to harsh mineral acids.