Energy 11 PROTECT Glovebox Solution

Energy 11 PROTECT Glovebox Solution

The Thermo Scientific Energy 11 PROTECT is a complete glovebox solution, combining twin-screw extrusion capabilities and a precisely controlled argon atmosphere within a user-friendly setup for easy handling.
Have Questions?
Catalog number 553-0800
Price (CLP)
-
Lithium-ion battery materials need to be processed under dry or even inert conditions to protect the sensitive materials involved. The Thermo Scientific Energy 11 PROTECT is a complete glovebox solution to this challenge, combining twin-screw extrusion capabilities and a precisely controlled argon atmosphere within a user-friendly setup for easy handling.

Due to its wide range of accessories, the Energy 11 PROTECT integrated glovebox solution allows users to develop and produce cathode and anode slurries as well as electrolytes for solid state batteries. Its high dispersive and distributive mixing capabilities also make possible the processing of dry electrodes under an argon atmosphere.

All extruder product contact parts can be disassembled completely and quickly for easy and thorough cleaning and to prevent cross contaminations. Rapid changes to experiment conditions are enabled by this feature as well.

Whatever your material requirements are, a range of feeding options for solid and liquid materials, all controlled by the extruder’s control panel, make sure that all ingredients go into the process precisely. Being a continuous process by design, the mixing done by the Energy 11 extruder eliminates batch to batch variations and always delivers a thoroughly homogenous final product.
 

APPLICATIONS

• All solid-state battery material mixing
• Handling of pure lithium anodes
• Separator film extrusion
• Lithium-ion electrode slurry mixing
• Solvent reduced mixing and dry cathode processing

Specifications
Amperage16
DescriptionEnergy 11 PROTECT Glovebox with intergated Energy 11 Twin-Screw Extruder
Dimensions (L x W x H)320 x 140 x 240 cm
For Use With (Application)Battery materials
Includes
  • Inert atmosphere with < 1ppm O2 and <1ppm H2O using argon (1-10 mbar overpressure)
  • Optional nitrogen removal  < 1 ppm N2 
  • Solvent filter for NMP removal in box atmosphere
Voltage400 V
Product TypeGlove Box
Unit SizeEach