This product will no longer be available on thermofisher.com after Dec. 28, 2020. Learn more »
Thermo Scientific™
Shandon™ Suresorb™ Clinical Pads and Formaldehyde Neutralizer Pads
Related applications:
Anatomical Pathology
Absorb and lock in liquids with Thermo Scientific™ Shandon™ Suresorb™ Clinical Pads and Formaldehyde Neutralizer Pads. Ideal for containing and disposing of body fluids containing Hepatitis B virus, AIDS / HIV and other pathogens.
Back to top
  • Fiber resists tearing, even when saturated
  • Formaldehyde neutralizer pads reduce exposure by chemically bonding with formaldehyde, resulting in the encapsulation of vapors—ideal for grossing areas and autopsy work
  • Clinical pads work as surgical cutting board pads, dissecting pads, autopsy table pads and liners for body trays and cadaver bags