HAAKE™ Rheomex OS Twin Screw Extruder for HAAKE™ PolyLab™ OS Torque Rheometer
Thermo Scientific™
HAAKE™ Rheomex OS Twin Screw Extruder for HAAKE™ PolyLab™ OS Torque Rheometer
With only a small sample quantity, simulate production processes, investigate rheological and substance-specific properties, and test and inspect new formulations with a HAAKE™ Rheomex OS Twin-Screw Extruder for the HAAKE™ PolyLab™ OS System torque rheometer.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoBarrel Bore DiameterBarrel Length L/D
567-750816 mm25 L/D
567-203131.8/20 mm (rear/front)
567-750416 mm40 L/D
567-752024 mm28 L/D
567-752124 mm40 L/D
Número do catálogo 567-7508
Preço (BRL)
-
Barrel Bore Diameter:
16 mm
Barrel Length L/D:
25 L/D
HAAKE™ Rheomex OS Twin-Screw Extruder for the HAAKE™ PolyLab™ OS Torque Rheometer System is available either as a parallel twin-screw extruder or a conical twin-screw extruder and attaches via a plug-and-play connection to the modular HAAKE Polylab Rheodrive OS. With a small sample quantity, customers can simulate production processes, investigate rheological and substance-specific properties, and test, and inspect new formulations.
The HAAKE Rheomex CTW 100 OS is a conical counter-rotating Twin-Screw Extruder that can be used for a wide range of tasks in laboratories and pilot plants when used together with specially designed screws with differing geometries. The conical twin-screw extruder is used for high-shear compounding of materials such as PVC, wood fiber compounding, and blending of polymers with different viscosities. A wide range of applications can be performed using different designed screw to adapt to the needs of the laboratory or pilot plant. The counter-rotating design supplies a positive material displacement from the hopper to the die, delivering high levels of pressure and shear, while allowing control of residence times.
The HAAKE Rheomex PTW 16 or 24 parallel twin-screw extruders have screw diameter of 16 mm or 24 mm, respectively. The horizontally split barrel, with the top half fitting upward, allows easy inspection of the melting and mixing. Parallel twin-screw, small-scale compounder are used for instance for running early-stage compounding and blending tests. For optimization of this behavior, the screws can be configured individually with elements for conveying, mixing and kneading. The segmented top barrel half is constructed in easy to configure modules for feeding and venting.
  • Continuous compounding with twin-screw extruders
  • Twin-screw laboratory extruders have a proven record in continuous compounding with feed-dosing of different additives (liquids or solids) along the extruder barrel
  • Combination of a twin-screw extruder unit with additional sensors offers the ability to measure material properties of the melt during processing
Typical twin-screw investigations:
  • Compounding of polymers with additives, reinforcing fibers, and fillers
  • Viscosity and flow behavior of the compound
  • Extrudability and scale-up data
  • Reactive extrusion
  • Morphology of polymer and nanocomposites
  • Decomposition of biopolymers
  • Influence on screw design on the viscosity
Compatible with:
Peripheral devices such as feeding systems, application-specific screws, mixer rotors, or extruder downstream equipment combine different modules to a complete downsized production line for the testing or small-scale production of new materials:
  • Extrudate cooling baths
  • Take-off units
  • Blown film tower
  • Inspection systems
  • Feeders
  • Pelletizers
  • Melt pumps
  • Capillary rheometers
Especificações
BarrelParallel, horizontally split
Barrel Bore Diameter16 mm
Barrel Zones7 - Heating/cooling zones
DescriptionRheomex PTW 16/25 OS Parallel Twin-Screw Extruder
IP RatingIP20
Max. Temperature 400 °C (optional 450 °C)
Barrel Length L/D25 L/D
Unit SizeEach