Nalgene™ Utility Boxes
Nalgene™ Utility Boxes
Nalgene™ Utility Boxes
Nalgene™ Utility Boxes
Thermo Scientific™

Nalgene™ Utility Boxes

Use these attractive, easy-to-clean boxes, available in four different sizes and shapes, for general-purpose laboratory storage.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호용량(미터법)제품 사이즈
5700-0500500 mLCase of 12
5700-0500PK
5700-0500으로도 사용됨
500 mLPack of 2
5700-10001000 mLCase of 12
5700-20002000 mLCase of 8
카탈로그 번호 5700-0500
제품 가격(KRW)
-
용량(미터법):
500 mL
Unit Size:
Case of 12

Use these attractive, easy-to-clean Thermo Scientific™ Nalgene™ Utility Boxes for general-purpose laboratory storage. Ideal for multi-purpose lab storage, boxes are available in four different sizes and shapes. Lids snap securely in place, clear walls offer easy view of contents and rounded corners allow for easy cleaning.

  • Manufactured from clear styrene-acrylonitrile with tight-fitting, molded, white, LDPE lids that include inserts to store and keep items dry and secure
  • Attractive and easy to clean
  • Stackable for high-density storage
  • Cat. No. 5700-0500 is ideal for electrophoresis gels
  • Not leakproof
사양
용량(영국식 단위)16 oz.
용량(미터법)500 mL
설명Sample Box SAN 500mL
치수(길이 x 폭 x 높이)7.5 x 6 x 1.5 in. (19 x 16 x 4 cm)
높이(영국식 단위)1.5 in.
길이(영국식 단위)7.5 in.
수량2/Pk.; 12/Cs.
폭(영국식 단위)6 in.
마개 유형Lid
색상Clear, White
높이(미터법)4 cm
길이(미터법)19 cm
폭(미터법)16 cm
Unit SizeCase of 12