Thermo Scientific™
Modular Filing System
 Related applications:
Anatomical Pathology
Hold a combination of microscope slides, tissue cassettes and 35mm slides in one cabinet with the Thermo Scientific™ Modular Filing System. Ideal for a variety of space and storage situations.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Modular System Features
  • Stackable unit made from anodized aluminum for years of dust-free storage
  • Available as a two-cell or five-cell cabinet
  • Five-cell cabinet stores up to 4600 microscope slides, 1500 of 35mm slides or 1000 tissue cassettes
Speculations
  • Five-Cell Cabinet Dimensions: 23.125 × 14.5 × 4 in. (58.7 × 36.8 × 10.2cm)
  • Microscope Slide Drawer and Tissue Cassette Drawer Dimensions: 24 × 3 × 4 in.(61 × 7.6 × 10.2cm)
  • Modular Base Dimensions: 25 × 15 × 4.5 in.(63.5 × 38.1 × 11.4cm)
Ordering Information
  • Drawers, base and slide separators are sold separately.
  • Foam dividers for slide drawers (Cat. No. B1000152) are available in packs of 12.
Documents
Manuals & protocols