HPLC Qualification Kits for UltiMate™ 3000 Systems
Actual product may vary
Thermo Scientific™
HPLC Qualification Kits for UltiMate™ 3000 Systems
HPLC Qualification Kits are for system validation of UltiMate 3000 LC and UHPLC systems.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호설명
5911.0050UCI-100 OQ Kit
3323.0010OP/PQ Standards Refill Kit
3321.0010AIQ Standards for Systems with UV Detector (Caffeine/Uracil)
4832.5000AQualification Kit (OQ/PQ) - Manual Injector
5705.0050AColumn Thermostat PQ Kit
4832.5010AQualification Kit (OQ/PQ) - Autosampler
3325.0010OQ/PQ Standards for RI-Detector
카탈로그 번호 5911.0050
제품 가격(KRW)
-
설명:
UCI-100 OQ Kit
Choose Thermo Scientific™ Dionex™ UltiMate™ HPLC Qualification Kits for system validation of UltiMate 3000 LC and UHPLC systems.
The modules of UltiMate 3000 RSLCnano system can be arranged in different configurations for various applications.
  • Components include fittings, capillaries and columns, but vary by application
사양
설명UCI-100 OQ Kit
용도(애플리케이션)Liquid Chromatography
용도 (장비)Dionex UltiMate 3000 Series
제품라인UltiMate
유형OQ Analysis Kit
Unit SizeEach