CaptureSelect™ C-tagXL MiniChrom™ Pre-packed Column
Thermo Scientific™

CaptureSelect™ C-tagXL MiniChrom™ Pre-packed Column

Thermo Scientific™ CaptureSelect™ C-tagXL MiniChrom™ columns are designed for bench-scale resin screening, process development, and sample preparation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColumn Capacity
59430720011 mL
59430720055 mL
Catalog number 5943072001
Price (USD)
620.00
Each
Column Capacity:
1 mL
Price (USD)
620.00
Each

CaptureSelect C-tagXL MiniChrom™ columns are designed for bench-scale resin screening, process development, and sample preparation.

These pre-packed columns are ready to connect and have the following features:

  • Dimensions of 1 mL (0.8 x 2 cm) and 5 mL (0.8 x 10 cm)
  • Direct connection to standard chromatography systems
  • Packed with CaptureSelect C-tagXL Affinity Matrix, offering high dynamic binding capacity and high resolution over a wide range of flow rates

The 1-mL columns can be used for quick screening of application feasibility and lab-scale purification on a convenient and easy-to-use pre-packed column. The 10-cm bed height of the 5-mL column allows initial process development on a bench scale.

CaptureSelect C-tagXL Affinity Matrix has a unique selectivity for a small 4-amino acid peptide tag (E-P-E-A, glutamic acid-proline-glutamic acid-alanine). CaptureSelect C-tagXL Affinity Matrix purifies the C-terminal tagged proteins with high affinity and selectivity, even in the presence of urea and guanidine HCl, from complex mixtures like cell culture harvests and periplasmic fractions in a one-step process. Mild elution conditions at neutral pH can be applied using magnesium chloride or propylene glycol, which ensures high-activity recoveries of pH-sensitive target proteins. The affinity resin recognizes the E-P-E-A tag sequence when fused either directly to the C-terminus of a protein or through a linker between the C-terminus and the E-P-E-A tag.

Features of this affinity matrix include:

  • Mild elution, making it suitable for pH-sensitive proteins
  • Binding of the tagged proteins under denaturing conditions (like dissolved inclusion bodies)
  • Excellent scalability
  • Non-animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose-based, epoxide-activated
  • Average particle size: 65 μm
  • Ligand: CaptureSelect C-tagXL affinity ligand
  • Ligand coupling method: epoxide coupling
  • Binding capacity: 400 nmol/mL resin depending on flow rate, column height, contact time
  • Elution conditions: Acidic: 20 mM citric acid or acetic acid, pH 3–4; 100 mM glycine buffer, pH 2–3. Neutral (pH 7.0–7.4): 20 mM Tris, 2.0 M MgCl2; 20 mM Tris, 1.0 M NaCl, 50% (v/v) propylene glycol; 20 mM Tris, 2 mM 'S-E-P-E-A' peptide
  • Column dimensions: 1 mL (0.8 x 2 cm) and 5 mL (0.8 x 10 cm)
  • Flow characteristics: 75–150 cm/h (up to 2 bar)
  • Formulation buffer: 20%(v/v) ethanol
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Bed Volume1 mL
Binding Capacity400 nmol/mL
Column Capacity1 mL
Column FormatPre-packed
Flow Rate75 to 150 cm/hr. (Up to 2 bar)
For Use With (Equipment)Chromatography Systems
Ligand TypeC-tagXL affinity ligand
MatrixAgarose
Particle Size65 μm
Quantity1 Column
Shipping ConditionWet Ice
Stationary PhaseC-tagXL
Diameter (Metric) Inner0.8 cm
Length (Metric)2 cm
Pore SizeNot Applicable
Product LineCaptureSelect
TypeAffinity Column
Unit SizeEach
Contents & Storage
Store at 4°C