CaptureSelect™ KappaXP MiniChrom™ Pre-packed columns
Thermo Scientific™

CaptureSelect™ KappaXP MiniChrom™ Pre-packed columns

The Thermo Scientific™ CaptureSelect™ KappaXP MiniChrom™ column is designed for bench-scale resin screening, process development, and sample preparation.
Have Questions?
Número de catálogoCantidad
59432120055 mL
Número de catálogo 5943212005
Precio (USD)
-
Cantidad:
5 mL

CaptureSelect KappaXP MiniChrom™ column is designed for bench-scale resin screening, process development, and sample preparation.

This pre-packed column is ready to connect and has the following features:

  • Volume of 5 mL (0.8 x 10 cm)
  • Direct connection to standard chromatography systems
  • Packed with CaptureSelect KappaXP Affinity Matrix, offering high dynamic binding capacity and high resolution over a wide range of flow rates

The 10-cm bed height of the 5-mL column allows initial process development on a bench scale.

CaptureSelect KappaXP matrix recognizes the kappa light chain of human antibodies, enabling purification of all Kappa containing Ig's, Fab, and Fab2 fragments directly from complex sources and feed streams.

Features of this affinity matrix include:

  • Purification of Kappa monoclonal antibodies, Fab, and Fab2 fragments in a single step with high purity and yield
  • Mild elution for Fab fragments and antibodies
  • Human-specific, no binding to bovine antibodies
  • Excellent scalability
  • Excellent pH stability up to 100 mM NaOH (limited exposure)
  • Non-animal derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics

  • Matrix: agarose-based, epoxide-activated
  • Average particle size: 65 μm
  • Ligand: CaptureSelect KappaXP affinity ligand
  • Ligand coupling method: epoxide coupling
  • Fab binding capacity: 20–30 g IgG Kappa-Fab/liter resin depending on flow rate, column height, and residence time
  • IgG binding capacity: 35–45 g IgG/liter resin depending on flow rate, column height
  • Elution conditions: 20 mM citric or acetic acid, pH 3–4; 100 mM Tris, 1.5 M MgCl2, pH 6
  • Column dimensions: 5 mL (0.8 x 10 cm)
  • Flow characteristics: 75–150 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Volumen del lecho5 mL
Formato de columnaPre-packed
Caudal75 to 150 cm/hr. (Up to 2 bar)
Para utilizar con (equipo)Chromatography Systems
Tipo de ligandoCaptureSelect KappaXP Affinity Ligand
MatrizAgarose
Presión máxima2 bar
Tamaño de partículas65 ±10 μm
Cantidad5 mL
Condiciones de envíoWet Ice
Fase estacionariaKappaXP Affinity Matrix with KappaXP Ligand
Diámetro interior (métrico)0.8 cm
Longitud (métrico)10 cm
Tamaño de poroNot Applicable
Línea de productosCaptureSelect MiniChrom
TipoPre-packed Column
Unit SizeEach
Contenido y almacenamiento
Store at 2°C to 8°C.