CaptureSelect™ KappaXP RoboColumn™ Pre-packed columns
Thermo Scientific™

CaptureSelect™ KappaXP RoboColumn™ Pre-packed columns

Thermo Scientific™ CaptureSelect™ RoboColumn™ are small chromatography columns used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.
Have Questions?
Número de catálogoCantidad
5943212200200 μL
Número de catálogo 5943212200
Precio (USD)
-
Cantidad:
200 μL

CaptureSelect RoboColumn™ are small chromatography columns used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.

CaptureSelect KappaXP matrix recognizes the kappa light-chain of human antibodies, enabling purification of all Kappa containing Ig's, Fab, and Fab2 fragments directly from complex sources and feed streams.

Features of this affinity matrix include

  • Purification of Kappa monoclonal antibodies, Fab, and Fab2 fragments in a single step with high purity and yield
  • Mild elution for Fab fragments and antibodies
  • Human-specific, no binding to bovine antibodies
  • Excellent scalability
  • Excellent pH stability up to 100 mM NaOH (limited exposure)
  • Non-animal derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Characteristics

  • Column size: 200 μL, 1 strip, 8 columns (8-row)
  • Column dimension: 5-mm diameter, 10-mm bed height
  • Matrix: agarose based, epoxide-activated
  • Average particle size: 65 μm
  • Ligand: CaptureSelect KappaXP affinity ligand
  • Ligand coupling method: epoxide coupling
  • Fab binding capacity: 20–30 g IgG Kappa-Fab/liter resin depending on flow rate, column height, and residence time
  • IgG binding capacity: 35–45 g IgG/liter resin depending on flow rate, column height, and residence time
  • Elution conditions: 20 mM citric or acetic acid, pH 3–4; 100 mM Tris, 1.5 M MgCl2, pH6
  • Formulation buffer: 20% (v/v) ethanol
  • Max operating pressure: 2 bar
  • Flow rate: 16–150 cm/hr

RoboColumn is a trademark of Repligen.

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.

Especificaciones
Volumen del lecho200 μL
Formato de columnaPre-packed Robo Column
Para utilizar con (equipo)Chromatography Systems
Tipo de ligandoCaptureSelect KappaXP Affinity Ligand
MatrizAgarose
Presión máxima2 bar
Tamaño de partículas65 ±10 μm
Cantidad200 μL
Condiciones de envíoWet Ice
Fase estacionariaKappaXP Affinity Matrix with KappaXP Ligand
Diámetro interior (métrico)0.5 cm
Longitud (métrico)1 cm
Tamaño de poroNot Applicable
Línea de productosCaptureSelect RoboColumn
Unit SizeEach
Contenido y almacenamiento
Store at 2°C to 8°C.