MabCaptureC™ RoboColumn™ Pre-packed Column
Thermo Scientific™

MabCaptureC™ RoboColumn™ Pre-packed Column

Thermo Scientific™ MabCaptureC™ RoboColumn™ are small chromatography columns used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberBed Volume
5943662600600 μL
5943662200200 μL
Catalog number 5943662600
Price (CLP)
867.653
Each
Bed Volume:
600 μL
Price (CLP)
867.653
Each

MabCaptureC™ RoboColumn™ are small chromatography columns used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.

These pre-packed columns are ready to connect and have the following features:

  • Packed with MabCaptureC™ Affinity Matrix, offering high dynamic binding capacity and high resolution over a wide range of flow rates
  • Direct connection to standard chromatography systems

The 1 mL columns can be used for quick screenings of application feasibility and lab scale purification on a convenient and easy-to-use pre-packed column. The 10-cm bed height of the 5 mL column allows initial process development at a bench scale.

The pre-packed MabCaptureC™ Affinity Matrix contains engineered rProtein A that binds at the CH2-CH3 interface of the Fc-region of IgGs and shows cross binding to the VH3 domain of IgGs.

Features of MabCaptureC™ Affinity Matrix include:

  • High dynamic mAb binding capacity of > 50 g/L at 4.8 min residence time
  • Purification of mAbs and a subpopulation of scFv's and Fab fragments containing VH3 binding domains
  • Clean and sanitize with standard sodium hydroxide agents for chromatography products
  • Excellent scalability
  • Non-animal derived

Main resin characteristics:

  • Matrix: agarose-based, epoxide activated
  • Average particle size: 75 ± 10 μm
  • Ligand: recombinant C domain of Protein A
  • Ligand coupling method: epoxide coupling
  • Binding capacity: >55 g IgG/L resin at 5 min residence time and 10% breakthrough; >75 g IgG/L resin at 10 min. residence time and 10% breakthrough
  • Elution conditions: 0.1 M glycine pH 3.0, or 20–100 mM sodium citrate or acetate, pH 3.0
  • Cleaning conditions: 0.2 M sodium hydroxide for 15 minutes every cycle
  • Column dimensions: 0.8 x 2 cm (1 mL), 0.8 x 10 cm (5 mL)
  • Operating pressure: ≤ 3 bar
  • Formulation buffer: 20% (v/v) ethanol
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Bed Volume600 μL
Binding Capacity>55 to 75 g IgG/L Resin at 10% breakthrough (depending on residence time)
Column FormatPre-packed RoboColumn
For Use With (Equipment)Robotic Fluid Handlers
Ligand TypeEngineered rProtein A
MatrixAgarose
Max. Pressure≤3 bar
Particle Size70 μm
Quantity1 Strip, 8 Columns
Shipping ConditionWet Ice
Stationary PhaseAgarose bead
TargetIgG mAbs, and a subpopulation of scFv and Fab containing VH3 domain
Diameter (Metric) Inner0.5 cm
Length (Metric)3 cm
Pore SizeNot Applicable
Product LineMabCaptureC
TypeProtein A Column
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.