CaptureSelect™ LambdaXP RoboColumn™ Pre-packed Columns
Thermo Scientific™

CaptureSelect™ LambdaXP RoboColumn™ Pre-packed Columns

CaptureSelect RoboColumns are small chromatography columns used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberBed Volume
5943752200200 μL
5943752600600 μL
Catalog number 5943752200
Price (MXN)
-
Bed Volume:
200 μL

CaptureSelect RoboColumn™ are small chromatography columns used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.

The CaptureSelect LambdaXP affinity matrix in this RoboColumn recognizes the constant domain of the human lambda light chain enabling purification of all IgG isotypes, including IgG, IgA, IgM, IgD, and IgE, directly from complex source materials in a robust and efficient purification process with excellent purity obtained in a single step.

Features of the LambdaXP affinity matrix include:

  • Efficient elution of Fab fragments and immunoglobulins containing a lambda light chain; no cross binding to the Fc-region or kappa light chains
  • One-step purification from crude material with high purity and yield
  • Human specific
  • Non-animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Characteristics

  • Column size: 200 μL, 1 strip, 8 columns (8-row)
  • Column dimension: 5-mm diameter, 10-mm bed height
  • Matrix: agarose-based, epoxide-activated
  • Average particle size: 65 μm
  • Ligand:CaptureSelect LambdaXP affinity ligand
  • Ligand coupling: epoxide coupling
  • Binding capacity: >30 g IgG/L resin depending on flow rate, column height, and contact time
  • Elution conditions: 25 mM sodium acetate, pH 3.5
  • Formulation buffer: 20% (v/v) ethanol
  • Operating pressure: 2 bar (maximum)
  • Flow rate: 16–150 cm/hr

RoboColumn is a trademark of Repligen.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Bed Volume200 μL
Column Capacity200 μL
Column FormatPre-packed RoboColumn
For Use With (Equipment)Robotic Fluid Handlers
Ligand TypeCaptureSelect LambdaXP Affinity Ligand
MatrixAgarose
Max. Pressure2 bar
Particle Size65 μm
Quantity1 Strip, 8 Columns
Shipping ConditionWet Ice
Diameter (Metric) Inner0.5 cm
Length (Metric)1 cm
Pore SizeNot Applicable
Product LineCaptureSelect
TypeAffinity Column
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C. Protect from light. Do not freeze.