Thermo Scientific™
Cytospin™ 4 Cytocentrifuge Accessories
 Related applications:
Anatomical Pathology
Use these accessories with the Thermo Scientific™ Cytospin™ 4 Cytocentrifuge.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Shorten preparation time by having a second sealed head ready when the first finishes processing
  • Autoclavable
  • 12-chamber head seals completely
Documents
Manuals & protocols