Samco™ Tube Closures
Samco™ Tube Closures
Samco™ Tube Closures
Samco™ Tube Closures
Thermo Scientific™
Samco™ Tube Closures
Depend on the reliable performance of these tube closures, available as flanged caps, two-position snap caps and universal caps.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberClosure SizeFor Use With (Equipment)
5A0008
also known as 5A 0008
17 mmPolypropylene Tubes
5A0007
also known as 5A 0007
17 mmPolystyrene Tubes
M99513312 to 16 mmTubes
Catalog number 5A0008
also known as 5A 0008
Price (MXN)
-
Closure Size:
17 mm
For Use With (Equipment):
Polypropylene Tubes
Depend on the reliable performance of Thermo Scientific™ Samco™ Tube Closures. Choose from two-position snap caps and universal caps. Our processes are monitored with quality checks which provide continued lot-to-lot uniformity and reliable performance.
Specifications
Closure Size17 mm
For Use With (Equipment)Polypropylene Tubes
No. per Case1000
No. per Pack200
Product LineSamco
ColorNatural
MaterialPolyethylene
Product Type2-position Scan Cap
SterilityNon-sterile
Unit SizeEach