Thermo Scientific™
Caps for Samco™ Histology Bio-Tite™ Nonsterile Specimen Containers
Prevent bio-hazardous spills with these Caps for Thermo Scientific™ Samco™  Histology Bio-Tite™  Specimen Containers.
Back to top
  • Bio-Tite caps create leak-resistant seal for superior safety and handling
  • Dual-threaded caps create improved sealing
Ordering Information:
Bulk packed, one bag per case