Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Prevent bio-hazardous spills with these Caps for Thermo Scientific™ Samco™ Histology Bio-Tite™ Specimen Containers.
Product Type | Specimen Container |
Closure Included | Yes |
Shape | Cylindrical |
Sterility | Non-sterile |
Closure Color | White |
Closure Size | 53 mm |
Closure Type | Bio-Tite Lid |
Description | Caps, 53mm; White |
Diameter (Metric) | 53 mm |
No. per Case | 1500/Cs. |
No. per Pack | 1500/Pk. |
Unit Size | Case of 1500 |
Product Type | Specimen Container |
Closure Included | Yes |
Shape | Cylindrical |
Sterility | Non-sterile |
Capacity (Metric) | 20 mL |
Closure Color | White |
Closure Size | 35 mm |
Closure Type | Bio-Tite Lid |
Description | Caps, 35mm; White |
Diameter (Metric) | 35 mm |
No. per Case | 4000/Cs. |
No. per Pack | 4000/Pk. |
Unit Size | Case of 4000 |
Product Type | Specimen Container |
Closure Included | Yes |
Shape | Cylindrical |
Sterility | Non-sterile |
Capacity (Metric) | 20 mL |
Closure Color | Green |
Closure Size | 35 mm |
Closure Type | Bio-Tite Lid |
Description | Caps, 35mm; Green |
Diameter (Metric) | 35 mm |
No. per Case | 4000/Cs. |
No. per Pack | 4000/Pk. |
Unit Size | Each |
Product Type | Specimen Container |
Closure Included | Yes |
Shape | Cylindrical |
Sterility | Non-sterile |
Closure Color | White |
Closure Size | 48 mm |
Closure Type | Bio-Tite Lid |
Description | Caps, 48mm; White |
Diameter (Metric) | 48 mm |
No. per Case | 2000/Cs. |
No. per Pack | 2000/Pk. |
Unit Size | Case of 2000 |
Product Type | Specimen Container |
Closure Included | Yes |
Shape | Cylindrical |
Sterility | Non-sterile |
Closure Color | Red |
Closure Size | 48 mm |
Closure Type | Bio-Tite Lid |
Description | Caps, 48mm; Red |
Diameter (Metric) | 48 mm |
No. per Case | 2000/Cs. |
No. per Pack | 2000/Pk. |
Unit Size | Each |
Product Type | Specimen Container |
Closure Included | Yes |
Shape | Cylindrical |
Sterility | Non-sterile |
Closure Color | Green |
Closure Size | 48 mm |
Closure Type | Bio-Tite Lid |
Description | Caps, 48mm; Green |
Diameter (Metric) | 48 mm |
No. per Case | 2000/Cs. |
No. per Pack | 2000/Pk. |
Unit Size | Each |
Product Type | Specimen Container |
Closure Included | Yes |
Shape | Cylindrical |
Sterility | Non-sterile |
Closure Color | Natural |
Closure Size | 48 mm |
Closure Type | Bio-Tite Lid |
Description | Caps, 48mm; Natural |
Diameter (Metric) | 48 mm |
No. per Case | 2000/Cs. |
No. per Pack | 2000/Pk. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Closure Size | Closure Color | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
5A0052 also known as 5A 0052 | Case of 1500 | 53 mm | White | Contact Us › | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
5A0032 also known as 5A 0032 | Case of 4000 | 35 mm | White | Contact Us › | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
5A0034 also known as 5A 0034 | Each | 35 mm | Green | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
5A0042 also known as 5A 0042 | Case of 2000 | 48 mm | White | Contact Us › | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
5A0043 also known as 5A 0043 | Each | 48 mm | Red | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
5A0044 also known as 5A 0044 | Each | 48 mm | Green | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
5A0050 also known as 5A 0050 | Each | 48 mm | Natural | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Ordering Information:
Bulk packed, one bag per case