HyperSep™ Pressure Manifold Waste Container
Thermo Scientific™
HyperSep™ Pressure Manifold Waste Container
Collect waste from the Thermo Scientific™ HyperSep™ Positive Pressure Manifold using this pre-drilled waste container.
Have Questions?
Catalog NumberFor Use With (Equipment)
60104-270Positive Pressure Manifold
Catalog number 60104-270
Price (USD)
1,512.36
Each
For Use With (Equipment):
Positive Pressure Manifold
Price (USD)
1,512.36
Each
Use the Thermo Scientific™ HyperSep™ Positive Pressure Manifold Waste Container to collect waste from the manifold. The HyperSep Positive Pressure Manifold holds up to 48 SPE cartridges. Samples and solvent, added to the SPE cartridge, are extracted under positive pressure for an even flow at each position. Extracts are collected into 13 or 16mm tubes.
Specifications
DescriptionPre-Drilled Waste Container, 1 pk
For Use With (Application)SPE
For Use With (Equipment)Positive Pressure Manifold
Product LineHyperSep
TypePre-Drilled Waste Container
Unit SizeEach