Empty Centrifuge Tubes for QuEChERS
Thermo Scientific™

Empty Centrifuge Tubes for QuEChERS

Empty Thermo Scientific™ Centrifuge Tubes for QuEChERS sample preparation are ideal for handling your QuEChERS samples. They come with screw caps for secure sealing, and are packaged separately.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)ColorQuantity
60106-CT-5050 mLBlue screw cap500
60106-CT-22 mLClear screw cap1000
60106-CT-1515 mLBlue screw cap500
Catalog number 60106-CT-50
Price (KRW)
-
Volume (Metric):
50 mL
Color:
Blue screw cap
Quantity:
500
Thermo Scientific™ Empty Centrifuge Tubes for QuEChERS sample preparation provide a reliable solution for your sample preparation needs. Featuring screw caps, these tubes help ensure secure sealing, and are designed for easy handling to fit your centrifugation needs.
  • Compatible with QuEChERS sample preparation
  • For use in centrifuges
  • Secure sealing with screw caps
  • Ideal for precise and efficient analysis of pesticides in foods

Specifications
Base MaterialPolypropylene
ColorBlue screw cap
DescriptionEmpty Centrifuge Tubes for QuEChERS
For Use With (Application)QuEChERS
Quantity500
TypeQuEChERS Sample Prep
Volume (Metric)50 mL
Unit SizePack of 500