HyperSep™ SPE Cartridges
Thermo Scientific™
HyperSep™ SPE Cartridges
Achieve greater reproducibility with cleaner more consistent extracts compared to conventional SPE, phospholipid removal and protein precipitation products.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSorbentType
60107-10-1-MCXMixed Mode Strong Cation Exchanger, PolymericSPE Cartridge 10 mg/1 mL
60107-30-1-HLBHLB (Reversed Phase Polymeric) SPE Cartridge 30 mg/1 mL
60107-30-1-MCXMixed Mode Strong Cation Exchanger, PolymericSPE Cartridge 30 mg/1 mL
60107-30-1-MAXMixed Mode Strong Anion Exchanger, PolymericSPE Cartridge 30 mg/1 mL
60107-10-1-HLBHLB (Reversed Phase Polymeric)SPE Cartridge 10 mg/1 mL
60107-10-1-MAXMixed Mode Strong Anion Exchanger, Polymeric SPE Cartridge 10 mg/1 mL
Catalog number 60107-10-1-MCX
Price (USD)
-
Sorbent:
Mixed Mode Strong Cation Exchanger, Polymeric
Type:
SPE Cartridge 10 mg/1 mL
Grow your laboratory productivity. Thermo Scientific™ HyperSep™ SPE Cartridges revolutionize solid phase extraction (SPE). This first fritless SPE product range enables greater sample throughput in bioanalytical laboratories where low elution volumes combined with greater reproducibility, lower failure rates and cleaner extracts are key to achieving better productivity.
HyperSep products are available in a 10 mg/1 mL cartridge format. They provide highly reproducible, clean and consistent sample extracts for bioanalysis by utilizing the unique award winning fritless SPE technology. The enhanced performance of HyperSep cartridges gives you greater confidence in your analytical results and lowers operating costs - without compromising ease of use or requiring complex method development.
HyperSep cartridges provide unparalleled performance characteristics compared to conventional SPE, phospholipid removal and protein precipitation products.
What HyperSep Cartridges Mean to Your Analysis
  • Higher levels of reproducibility
  • Higher levels of extract cleanliness
  • Reduced solvent requirements
  • Confidence in results
Specifications
For Use With (Application)Bioanalytical Applications - Strong Ion-Exchange Retention of Weak Basic Compounds. Complementary Reversed Phase Retention of Neutral Compounds.
Quantity100/pk
Bed Weight30 mg
Product LineHyperSep
SorbentMixed Mode Strong Cation Exchanger, Polymeric
TypeSPE Cartridge 10 mg/1 mL
Unit SizePack of 100