HyperSep™ PFAS Cartridges
Actual product may vary

HyperSep™ PFAS Cartridges

HyperSep SPE PFAS SPE products are designed to address the needs of extraction and concentration of PFAS samples for targeted and not targeted screening while minimizing background contamination.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberBed WeightParticle SizeSorbent
60107-200-6-WAX
also known as ECWAX126-P
200 mg55 to 65 μmPolymeric WAX
60107-150-6-WAX
also known as ECWAX(150)6-P
150 mg55 to 65 μmPolymeric WAX
60107-20050-WAXGCB
also known as ECWAXCB206-P
250 mg55 to 65 μm and 120/400 mesh200 mg WAX, 50 MG GCB
60107-50200-GCBWAX250 mg55 to 65 μm and 120/400 mesh50 mg GCB, 200 MG WAX
60107-500-6-HLD
also known as ECHLD156-P
500 mg20 to 40 μmPolymeric DVB
60107-500-6-WAX
also known as ECWAX156-P
500 mg55 to 65 μmPolymeric WAX
60107-30-3-WAX
also known as ECWAX063-P
30 mg55 to 65 μmPolymeric WAX
GCB-10
also known as EUCAB00X
10 g55 to 65 μm and 120/400 meshGraphitized Carbon Black (GCB)
Catalog number 60107-200-6-WAX
also known as ECWAX126-P
HSN 84212900
Price (INR)
29,401.00
Pack
Bed Weight:
200 mg
Particle Size:
55 to 65 μm
Sorbent:
Polymeric WAX
Price (INR)
29,401.00
Pack
HyperSep SPE PFAS SPE products are designed to address the needs of extraction and concentration of PFAS samples for targeted and not targeted screening while minimizing background contamination. The range of selectivity’s and formats are designed to meet the requirements for main environmental testing methods by extracting and concentrating samples from drinking water, ground water, surface water, wastewater, soil, food and other complex matrices.

Per- and polyfluorinated alkyl substances (PFAS) are among the primary emerging contaminants of concern. The detection and quantification of known PFAS and the discovery of unknown PFAS substances is rapidly growing. SPE solutions are a fundamental part of the analytical workflow for analysis of PFAS compounds.

Thermo Scientific provides a range of products to address PFAS testing requirements e.g. EPA methods: 1633, 537.1, 533.

Specifications
DescriptionHyperSep PFAS SPE, 200 mg, 6 mL, WAX
Bed Weight200 mg
For Use With (Application)EPA method 1633 (alternative), extraction of PFAS from water, tissue, soil, or biosolids
For Use With (Equipment)Vacuum Manifolds or Automated SPE Equipment
Particle Size55 to 65 μm
Product LineHyperSep
SorbentPolymeric WAX
TypeSolid Phase Extraction Column
Volume (Metric)6 mL
Quantity30/Pk.
Unit SizePack