HyperSep™ PFAS Cartridges
Thermo Scientific™

HyperSep™ PFAS Cartridges

HyperSep SPE PFAS SPE products are designed to address the needs of extraction and concentration of PFAS samples for targeted and not targeted screening while minimizing background contamination.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberBed WeightParticle SizeSorbent
60107-30-3-WAX30 mg55 to 65 μmPolymeric WAX
GCB-1010 g55 to 65 μm and 120/400 meshGraphitized Carbon Black (GCB)
Catalog number 60107-30-3-WAX
Price (TWD)
-
Bed Weight:
30 mg
Particle Size:
55 to 65 μm
Sorbent:
Polymeric WAX
HyperSep SPE PFAS SPE products are designed to address the needs of extraction and concentration of PFAS samples for targeted and not targeted screening while minimizing background contamination. The range of selectivity’s and formats are designed to meet the requirements for main environmental testing methods by extracting and concentrating samples from drinking water, ground water, surface water, wastewater, soil, food and other complex matrices.

Per- and polyfluorinated alkyl substances (PFAS) are among the primary emerging contaminants of concern. The detection and quantification of known PFAS and the discovery of unknown PFAS substances is rapidly growing. SPE solutions are a fundamental part of the analytical workflow for analysis of PFAS compounds.

Thermo Scientific provides a range of products to address PFAS testing requirements e.g. EPA methods: 1633, 537.1, 533.

Specifications
DescriptionHyperSep PFAS SPE, 30 mg, 3 mL, WAX
For Use With (Application)EU and other regulations, extraction of PFAS from water
Quantity30/Pk.
Bed Weight30 mg
For Use With (Equipment)Vacuum Manifolds or Automated SPE Equipment
Particle Size55 to 65 μm
Product LineHyperSep
SorbentPolymeric WAX
TypeSolid Phase Extraction Column
Volume (Metric)3 mL
Unit SizePack of 30