HyperSep™ C8 Cartridges
Thermo Scientific™

HyperSep™ C8 Cartridges

Need a less retentive alternative to C18 for polar and nonpolar compounds? These hydrophobic reversed phase C8 cartridges offer excellent performance.
Catalog NumberBed WeightColumn Capacity (Metric)
60108-4271000 mg6 mL
60108-309500 mg3 mL
60108-39150 mg1 mL
60108-392100 mg1 mL
60108-393200 mg3 mL
60108-394500 mg6 mL
60108-7042000 mg15 mL
60108-70610 g75 mL
Catalog number 60108-427
Price (JPY)
11,460
Online offer
Ends: 26-Jun-2026
19,100
Save 7,640 (40%)
Each
Bed Weight:
1000 mg
Column Capacity (Metric):
6 mL

The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.

Thermo Scientific™ HyperSep™ C8 Cartridges offer a less retentive alternative to C18 for polar and nonpolar compounds, and are well-suited for methods requiring less retention than C18. The cartridges are ideally suited for the analysis of:

  • Drugs and their metabolites in biological samples
  • Peptides in biological samples
HyperSep C8 SPE Cartridges provide excellent performance characteristics for moderately polar analytes. They also offer the potential for polar interactions greater than with C18.

HyperSep SPE Cartridges and Plates

HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.

The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column Capacity (Metric)6 mL
Description1 g Bed Weight; 6 mL Column Volume
For Use With (Application)Screening of Drugs, Metabolites, Peptides in Biological Samples
QuantityPack of 30
Bed Weight1000 mg
For Use With (Equipment)Thermo Scientific HyperSep-96 System
Particle Size40 to 60 μm
Product LineHyperSep
SorbentHyperSep C8
TypeSolid Phase Extraction Cartridge
Volume (Metric)6 mL
Unit SizeEach