HyperSep™ SAX Cartridges
Thermo Scientific™

HyperSep™ SAX Cartridges

Obtain excellent performance for extraction of carboxylic and other weak acids with these strong anion exchanger SPE cartridges.
Catalog NumberBed WeightColumn Capacity (Metric)
60108-7145000 mg25 mL
60108-360500 mg6 mL
60108-41750 mg1 mL
60108-418100 mg1 mL
60108-419200 mg3 mL
60108-4341000 mg6 mL
60108-521500 mg3 mL
60108-7132000 mg15 mL
60108-71510 g75 mL
Catalog number 60108-714
Price (JPY)
39,000
Each
Bed Weight:
5000 mg
Column Capacity (Metric):
25 mL
Price (JPY)
39,000
Each
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.

Thermo Scientific™ HyperSep™ SAX Cartridges feature a strong anion exchange sorbent for extraction of weak acids. Typical applications include:

  • Isolation of anionic proteins
  • Removal of acidic food pigments
  • Isolation of phenolic compounds
  • Nucleic acids, nucleotides and surfactants
HyperSep SAX Strong Anion Exchanger Cartridges are ideally suited for the extraction of carboxylic and other weak acids. The chemistry extracts negatively charged compounds from both aqueous and nonaqueous solutions.
  • Amine group masks the effect of the carbon chain of the functional group
  • Selectivity can be tuned by choice of buffer

HyperSep SPE Cartridges and Plates

Thermo Scientific™ HyperSep™ solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.

The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column Capacity (Metric)25 mL
Description5 g Bed Weight; 25 mL Column Volume
For Use With (Application)Extraction of Weak Acids
QuantityPack of 20
Bed Weight5000 mg
For Use With (Equipment)Thermo Scientific HyperSep-96 System
Particle Size40 to 60 μm
Product LineHyperSep
SorbentHyperSep SAX
TypeSolid Phase Extraction Cartridge
Volume (Metric)25 mL
Unit SizeEach