SMART Digest™ Trypsin Kits
Thermo Scientific™

SMART Digest™ Trypsin Kits

Obtain fast, simple, and highly reproducible trypsin protein digestion for peptide characterization and quantitation in biopharmaceutical protein research.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberFormatTypeIncludes
60109-103-MBBulk Magnetic ResinTrypsin KitResin, Buffer, Collection Plate, SOLAμ SPE Plate
60109-101Resin in Pre-filled PCR TubesTrypsin KitResin, Collection Plate
60109-101-LPHResin in Pre-filled PCR TubesTrypsin KitLow pH Resin, Collection Plate
60109-103Resin in Pre-filled PCR TubesTrypsin KitResin, Collection Plate, SOLAμ SPE Plate
60109-102Resin in Pre-filled PCR TubesTrypsin KitResin, Collection Plate, Filter Plate
60109-101-BBulk Non-magnetic ResinTrypsin KitResin, Buffer
60109-101-MBBulk Magnetic ResinTrypsin KitResin, Buffer
60109-102-BBulk Non-magnetic ResinTrypsin KitResin, Buffer, Collection Plate, Filter Plate
60109-102-MBBulk Magnetic ResinTrypsin KitResin, Buffer, Collection Plate, Filter Plate
60109-103-BBulk Non-magnetic ResinTrypsin KitResin, Buffer, Collection Plate, SOLAμ SPE Plate
60113-101Soluble TrypsinSoluble Trypsin KitSoluble Trypsin, Buffer, Collection Plate
60113-101-10PKSoluble TrypsinSoluble Trypsin Kit - 10 Pk.Soluble Trypsin, Buffer, Collection Plate
Catalog number 60109-103-MB
Price (USD)
1,287.97
Each
Format:
Bulk Magnetic Resin
Type:
Trypsin Kit
Includes:
Resin, Buffer, Collection Plate, SOLAμ SPE Plate
Price (USD)
1,287.97
Each

Obtain high quality analytical results from protein digests using Thermo Scientific™ SMART Digest™ Kits. A significant advance in sample preparation for biopharmaceutical protein research, the kits provide fast and simple protein digestion with high reproducibility, high sensitivity, and high levels of data quality in a format that's compatible with automation. The SMART Digest Kits are a significant improvement over current in-solution protein digestion technologies, which are not reproducible, have poor sensitivity, and employ protracted methodologies not amenable to automation.

Trypsin Protein Digestion

SMART Digest kits enable you to characterize and quantitate proteins faster, easier, and quicker than ever before. This version of the kit uses trypsin, which cleaves peptides on the C-terminal side of lysine and arginine amino acid residues.

Easy and simple

  • Significantly increased speed of sample processing
  • Simple process
  • Easily automate entire protocol for high-throughput processing

Reliable Results

  • Increased reproducibility over existing protocols
  • Higher sensitivity
  • Higher data confidence

Significantly Reduces Sample Preparation Time

Typically it takes less than 60 minutes (dependent on sample complexity) to achieve full digestion, whereas an in-solution digest can take up to 24 hours.

Cleaner Samples

Reduction in the number of chemicals used and the immobilized trypsin design helps mitigate against chemically induced post translational modifications (PTMs) and autolysis. This results in results which are easier to interpret.

Couple with SOLAμ SPE Plates for Desalting

When used with the SMART Digest kits for desalting, Thermo Scientific™ SOLAμ™ SPE solid phase extraction plates provide a fast, easy-to-use, robust, and easily automatable sample preparation solution for analysis of biomolecules.

Specifications
DescriptionSMART Digest Trypsin Kit Magnetic Bulk Resin option with SOLAμ/collection plate
For Use With (Application)Peptide Mapping
IncludesResin, Buffer, Collection Plate, SOLAμ SPE Plate
CaptureNone
CleavageC-terminal side of lysine and arginine amino acids
EnzymeTrypsin
FormatBulk Magnetic Resin
Product LineSMART Digest
TypeTrypsin Kit
Unit SizeEach