Thermo Scientific™

HyperSep™ Polystyrene Lab Plates

Catalog number: 60110-201
Thermo Scientific™

HyperSep™ Polystyrene Lab Plates

Catalog number: 60110-201
Available in a range of chromatographic materials, Thermo Scientific™ HyperSep™ Polystyrene Lab Plates enable simple, convenient purification and sample preparation for proteins, DNA, RNA and other biomolecules. In this 96 well plate format, media is embedded at the bottom of the plate. Typical applications include: 
  • Tissue culture and separation of products
  • Concentration of small-scale samples
  • Sample clean-up
  • Collection of sample after chromatography
 
Catalog Number
60110-201
Unit Size
Each
Description
C18; Polystyrene
Price (USD)
Availability
***
Full specifications
Quantity
-
DescriptionC18; Polystyrene
For Use With (Application)Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules
Particle Size40 to 60 μm
Product LineHyperSep
SorbentHyperSep C18
Type96-Well Plate
Unit SizeEach
Catalog Number
Unit Size
Pack of 5
Description
Zirconium Dioxide; Polystyrene
Price (USD)
474.60
Your Price:
Availability
***
Full specifications
Quantity
DescriptionZirconium Dioxide; Polystyrene
For Use With (Application)Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules
Product LineHyperSep
SorbentHyperSep Zirconium Dioxide
Type96-Well Plate
Unit SizePack of 5
Catalog Number
Unit Size
Pack of 5
Description
Titanium Dioxide; Polystyrene
Price (USD)
474.60
Your Price:
Availability
***
Full specifications
Quantity
DescriptionTitanium Dioxide; Polystyrene
For Use With (Application)Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules
Product LineHyperSep
SorbentHyperSep Titanium Dioxide
Type96-Well Plate
Unit SizePack of 5
Catalog Number
Unit Size
Pack of 5
Description
SAX; Polystyrene
Price (USD)
452.00
Your Price:
Availability
***
Full specifications
Quantity
DescriptionSAX; Polystyrene
For Use With (Application)Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules
Particle Size40 to 60 μm
Product LineHyperSep
SorbentHyperSep SAX
Type96-Well Plate
Unit SizePack of 5
Showing 4 of 4
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)AvailabilityQuantity
60110-201Full specifications
EachC18; PolystyreneRequest A Quote***-
DescriptionC18; Polystyrene
For Use With (Application)Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules
Particle Size40 to 60 μm
Product LineHyperSep
SorbentHyperSep C18
Type96-Well Plate
Unit SizeEach
60110-206Full specifications
Pack of 5Zirconium Dioxide; Polystyrene
474.60
Your Price:
***
DescriptionZirconium Dioxide; Polystyrene
For Use With (Application)Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules
Product LineHyperSep
SorbentHyperSep Zirconium Dioxide
Type96-Well Plate
Unit SizePack of 5
60110-207Full specifications
Pack of 5Titanium Dioxide; Polystyrene
474.60
Your Price:
***
DescriptionTitanium Dioxide; Polystyrene
For Use With (Application)Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules
Product LineHyperSep
SorbentHyperSep Titanium Dioxide
Type96-Well Plate
Unit SizePack of 5
60110-209Full specifications
Pack of 5SAX; Polystyrene
452.00
Your Price:
***
DescriptionSAX; Polystyrene
For Use With (Application)Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules
Particle Size40 to 60 μm
Product LineHyperSep
SorbentHyperSep SAX
Type96-Well Plate
Unit SizePack of 5
Showing 4 of 4
The convenient 96-well HyperSep Lab Plates can be processed manually or using a liquid-handling robot. They are not suitable for use with a vacuum.

Figures