SMART Digest Trypsin Kit, Soluble, 250 ug/250 uL Trypsin, 20 mL Digestion Buffer
Thermo Scientific™
SMART Digest Trypsin Kit, Soluble, 250 ug/250 uL Trypsin, 20 mL Digestion Buffer
Obtain fast, simple, and highly reproducible trypsin protein digestion for peptide characterization and quantitation in biopharmaceutical protein research.
Have Questions?
카탈로그 번호Cleavage
60113-250-10PKC-terminal side of lysine and arginine amino acids
카탈로그 번호 60113-250-10PK
제품 가격(KRW)
-
Cleavage:
C-terminal side of lysine and arginine amino acids
Obtain high quality analytical results for protein digests using Thermo Scientific™ SMART Digest™ Kits. A significant advance in sample preparation for biopharmaceutical protein research and quantitation, the kits provide fast and simple protein digestion with high reproducibility, high sensitivity, and high levels of data quality in a format that's compatible with automation. The SMART Digest Kits are a significant improvement over current in-solution protein digestion technologies, which are less reproducible and employ protracted methodologies not amenable to automation.
Trypsin Protein Digestion
SMART Digest kits enable you to characterize and quantitate proteins faster, easier, and quicker than ever before. This version of the kit uses trypsin, which cleaves peptides on the C-terminal side of lysine and arginine amino acid residues.
Easy and simple
  • Significantly increased speed of sample processing
  • Simple process
  • Easily automate entire protocol for high-throughput processing
Reliable Results
  • Increased reproducibility over existing protocols
  • Higher sensitivity
  • Higher data confidence
Significantly Reduces Sample Preparation Time
Typically it takes less than 60 minutes (dependent on sample complexity) to achieve full digestion, whereas an in-solution digest can take up to 24 hours.
Cleaner Samples
Reduction in the number of chemicals used and the immobilized trypsin design helps mitigate against chemically induced post translational modifications (PTMs) and autolysis. This results in results which are easier to interpret.
Couple with SOLAμ SPE Plates for Desalting
When used with the SMART Digest kits for desalting, Thermo Scientific™ SOLAμ™ SPE solid phase extraction plates provide a fast, easy-to-use, robust, and easily automatable sample preparation solution for analysis of biomolecules.
사양
용도(애플리케이션)Peptide Quantitation, Peptide Mapping
포함SMART Digest Soluble Trypsin, SMART Digest Buffer, Collection Plate
CleavageC-terminal side of lysine and arginine amino acids
효소Trypsin
형식SMART Digest Soluble Trypsin
제품라인SMART Digest
유형Protein Digestion
Unit SizePack of 10