SMART Digest RNase Columns
Thermo Scientific™

SMART Digest RNase Columns

Thermo Scientific SMART Digest™ RNase Columns provide simple, fast and highly reproducible digestion of ribonucleic acid (RNA) to enable oligonucleotide mapping and quantitation.

Catalog NumberEnzyme
60140-102RNase A
60140-101RNase T1
Catalog number 60140-102
Price (JPY)
264,780
Online offer
Ends: 26-Jun-2026
441,300
Save 176,520 (40%)
Each
Enzyme:
RNase A

Thermo Scientific SMART Digest Immobilized Ribonucleases (Rnases) Columns are designed using the same proven technology as the Thermo Scientific SMART Digest Rnase kits to provide simple, fast and highly reproducible digestion of ribonucleic acid (RNA) in a flow through format.

Obtain high-quality analytical results from mRNA digestions using SMART Digest RNase columns. A significant advance in sample preparation for oligonucleotide mapping, the columns provide fast, simple digestion with high levels of reproducibility, RNase columns use immobilized ribonucleases which allow control of partial digestion to produce required fragments of ideal length for sequencing by HRAM-MS.

Easy and simple

  • Simple process allows control of digestion time ensuring required fragment length for comprehensive mRNA sequencing by HRAM MS
  • Single nuclease digestion allows full characterization of mRNA

Reliable results

  • Increased reproducibility over existing protocols
  • Higher data confidence
  • No nuclease contamination of the analytical column

RNase columns are available with two enzyme specificities:

  • RNase T1 is an endoribonuclease that specifically degrades single-stranded RNA at G residues
  • RNase A is an endoribonuclease that specifically degrades single-stranded RNA at C and U residues
Specifications
Column FormatImmobilized enzyme
Column TypeImmobilized enzyme in PEEK cartridge
DescriptionSMART Digest RNase A column
Diameter2.1 mm
Flow Rate10 to 500 μL/min
For Use With (Application)Oligionucleotide Mapping
Length30 mm
Max. Pressure2500 psi (170 bar)
Mobile Phase CompatibilityAmmonium acetate or Tris without saline
Operating Temperature Range20 °C to 50 °C
Packing MaterialHydrophilic/coated DVB
Particle Size˜20 μm
Pore Size> 1000Å
Stability3 years
Stationary PhaseImmobilized enzyme
Storage Temperature4 °C for extended period of time (refrigerate in Tris without Saline)
TechniqueBioLC
Weight8 g
CleavageDegrades single-stranded RNA at C & U residues
EnzymeRNase A
TypeSMART Digest RNase A column
pH7 to 8.5
Unit SizeEach