Super Clean™ Gas Cartridge Filter Baseplates and Accessories
Super Clean™ Gas Cartridge Filter Baseplates and Accessories
Select from a range of quality baseplates/manifolds for your Thermo Scientific™ Super Clean™ Cartridge Filters.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
60180-814Single Base
60180-815Dual Base
60180-817Triple Base
60180-818Four-Position Base
Catalog number 60180-814
Price (USD)
-
Type:
Single Base
Use quality baseplates/manifolds for your Thermo Scientific™ Super Clean™ Cartridge Filters. A range of baseplate options allows configuration to your individual requirements. Needle valves ensure that the gas line is not contaminated during cartridge change. The baseplates have a one-time installation procedure. Super Clean Cartridge Filters are not included.
Additional Product Information
  • Use with Super Clean cartridge filters
  • Filters not included
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionFor Gas: Carrier Gas Only
For Use With (Equipment)Triple Filter
TypeSingle Base
Unit SizeEach