Search
| 產品號碼 | Barcode | No. per Pack |
|---|---|---|
| 60180-P341B | Includes Barcode | 100 |
| 60180-P340 | No Barcode | 10 Plates |
| 60180-P341 | No Barcode | 100 Plates |
| 60180-P340B | Includes Barcode | 10 Plates |
SureSTART Level 3 WebSeal Plate+ 384-Well Square-Rounded V-Bottom Microtiter Plates are designed for regulated labs that require high performing products.
Plate+ glass-coated Level 3 well plates are recommended for use with very sensitive applications, low sample concentrations, and hydrophobic samples such as proteins and are ideal for use with all solvent types including non-polar solvents.
These 384-well plates are specification certified to ensure autosampler compatible physical dimensions and conform to ANSI-SLAS 4-2004 standards. Seal these well plates with SureSTART WebSeal 384-Well Plate Sealing Mats, Square, Level 1 or SureSTART WebSeal 384-Well Plate Sealing Mats, Square, Level 2.
Performance Level 3 Vials – for High Performance Analysis
Specification Certified
Lot Traceable
About Thermo Scientific WebSeal Well Plates and Mats
Thermo Scientific WebSeal plates and mats are available in three performance levels: