Thermo Scientific™
HM Microtome Instrumentation Accessories
Related applications:
Anatomical Pathology
Choose from multiple Thermo Scientific™ HM Microtome Instrumentation Accessories for use with Thermo Scientific™ HM Microtomes.
Back to top
Automatic approach system:
  • Direct sectioning possible after the automatic approach
  • Adaptable to HM 335 E, HM 340 E, HM 355 S, and HM 360 microtomes
Backlighting system:
  • Illumination for block/knife relationship provided
  • Insertable into disposable blade carrier, knife carrier C, Standard, and S knife carrier
  • Ideal for semi-thin sectioning techniques
  • Useful for microscopic evaluation of the light reflex of the knife edge on the specimen