HyperSep™ Retain CX Plates
Thermo Scientific™

HyperSep™ Retain CX Plates

Achieve consistent, high recoveries for basic compounds with these Thermo Scientific™ HyperSep™ Retain CX Plates.
Have Questions?
Catalog NumberBed WeightVolume (Metric) Well
60306-30460 mg1 mL
Catalog number 60306-304
Price (USD)
496.56
Each
Bed Weight:
60 mg
Volume (Metric) Well:
1 mL
Price (USD)
496.56
Each
Achieve consistent, high recoveries for basic compounds with these Thermo Scientific™ HyperSep™ Retain CX Plates, packed with a high capacity, high purity, highly porous polystyrene DVB material partially modified with sulfonic acid functional groups to give superior recoveries for basic analytes. Used in SPE sample preparation, these cation exchange well plates and individual wells can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Widely used in forensic and clinical analysis of a wide range of drugs of abuse, including basic and neutral drugs. Hypersep Retain SPE wells conform to industry standard configurations and allow efficient routine processing by vacuum, positive pressure or centrifugation.
  • High, consistent recoveries
  • Ideal for large samples
  • Fast and easy sample preparation and method development
  • Reproducible, reliable sample preparation in a traditional format
  • pH stable 0 to 14
  • Available in a wide range of chemistries and bed weights
For Use With: The Thermo Scientific™ HyperSep-96™ System.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionRetain CX 60 mg/2 mL 96 Fixed Well Plate
For Use With (Application)Retention of Basic Analytes
Quantity1 Pk.
Volume (Metric) Well1 mL
Bed Weight60 mg
Particle Size30 to 50 μm
Product LineHyperSep
SorbentHyperSep Sulfonic Acid-Modified Polystyrene DVB
TypeSolid Phase Extraction Well Plate
Unit SizeEach