Viper™ TQ Capillary Kit for Vanquish Duo HPLC Systems
Thermo Scientific™
Viper™ TQ Capillary Kit for Vanquish Duo HPLC Systems
Boost your productivity and confidence, and accelerate method development using application-specific LC workflows.
Have Questions?
Número de catálogoIncluyePara utilizar con (equipo)
6036.2305AViper TQ Capillary Kit for Vanquish Duo UHPLC SystemsVanquish Duo UHPLC Systems
Número de catálogo 6036.2305A
Precio (CLP)
-
Incluye:
Viper TQ Capillary Kit for Vanquish Duo UHPLC Systems
Para utilizar con (equipo):
Vanquish Duo UHPLC Systems
Improve chromatographic resolution by minimizing dead volume with the Thermo Scientific™ Viper™ TQ UHPLC Fittings. These pre-assembled kits allow you to quickly connect LC modules, valves, and columns for leak-free fluidic HPLC and UHPLC connections. Ideally suited for running application-specific workflows with your HPLC and LC-MS systems, like the Thermo Scientific™ Vanquish™ Method Development LC Systems, Thermo Scientific Vanquish Online 2D-LC Systems, or Thermo Scientific Vanquish Duo UHPLC Systems.
Enjoy the confidence of robust and reliable fingertight LC connections giving you tactile and audible feedback. Tailored to your advanced workflow requirements, Viper TQ application-specific capillary kits offer:
  • Simplified fluidic connection for all user levels with torque-controlled fingertight connections with sensory feedback
  • Easy-to-install, tool-free reusable capillaries requiring up to 40% less force compared to non-torque controlled fittings
  • Reliable, reproducible LC connections that deliver low carryover and simplify method transfer
Especificaciones
IncluyeViper TQ Capillary Kit for Vanquish Duo UHPLC Systems
MaterialMP35N
Presión máxima1517 bar
Para utilizar con (equipo)Vanquish Duo UHPLC Systems
Línea de productosViper TQ
TipoCapillary Kit
Unit SizeEach